Assay Details
| Gene Symbol: LOC100355311 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717114.3 | XP_002717160.1 | 5 - 6 | 917 | 81 | |
| XM_008267761.2 | XP_008265983.1 | 5 - 6 | 920 | 81 | ||
More Information
Set Membership: |
|

LOC100355311
Genomic Map