Assay Details
| Gene Symbol: SEC61A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717524.3 | XP_002717570.1 | 6 - 7 | 596 | 67 | |
| XM_017347676.1 | XP_017203165.1 | 7 - 8 | 683 | 67 | ||
| XM_017347677.1 | XP_017203166.1 | 7 - 8 | 694 | 67 | ||
| XM_017347678.1 | XP_017203167.1 | 6 - 7 | 840 | 67 | ||
More Information
Set Membership: |
|

SEC61A2
Genomic Map