Assay Details
| Gene Symbol: SLC45A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714118.2 | XP_002714164.1 | 3 | 1014 | 85 | |
| XM_017344668.1 | XP_017200157.1 | 2 | 846 | 85 | ||
More Information
Set Membership: |
|

SLC45A2
Genomic Map