Assay Details
| Gene Symbol: PRKN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017345477.1 | XP_017200966.1 | 13 - 14 | 1308 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRKN| Gene Symbol: PRKN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017345477.1 | XP_017200966.1 | 13 - 14 | 1308 | 64 | |
Set Membership: |
|