Assay Details
| Gene Symbol: PXYLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716582.3 | XP_002716628.1 | 5 - 6 | 633 | 72 | |
| XM_008266253.2 | XP_008264475.1 | 4 - 5 | 670 | 72 | ||
| XM_017346813.1 | XP_017202302.1 | 5 - 6 | 644 | 72 | ||
More Information
Set Membership: |
|

PXYLP1
Genomic Map