Assay Details
| Gene Symbol: PTP4A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002724361.2 | XP_002724407.1 | 1092 | 55 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PTP4A3| Gene Symbol: PTP4A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002724361.2 | XP_002724407.1 | 1092 | 55 | ||
Set Membership: |
|