Assay Details
| Gene Symbol: GPATCH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709921.3 | XP_002709967.1 | 8 - 9 | 840 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPATCH11| Gene Symbol: GPATCH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709921.3 | XP_002709967.1 | 8 - 9 | 840 | 73 | |
Set Membership: |
|