Assay Details
| Gene Symbol: PAM16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249597.2 | XP_008247819.1 | 110 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008249597.2 transcripts.
Set Membership: |
|

PAM16
Genomic Map