Assay Details
| Gene Symbol: RAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260421.2 | XP_008258643.1 | 4 - 5 | 754 | 53 | |
| XM_008260422.2 | XP_008258644.1 | 3 - 4 | 641 | 53 | ||
More Information
Set Membership: |
|

RAF1
Genomic Map