Assay Details
| Gene Symbol: MAP3K20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008258754.2 | XP_008256976.1 | 5 - 6 | 669 | 79 | |
| XM_008258756.2 | XP_008256978.1 | 5 - 6 | 780 | 79 | ||
| XM_008258757.2 | XP_008256979.2 | 5 - 6 | 556 | 79 | ||
| XM_017342939.1 | XP_017198428.1 | 5 - 6 | 669 | 79 | ||
More Information
Set Membership: |
|

MAP3K20
Genomic Map