Assay Details
| Gene Symbol: SKP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710210.3 | XP_002710256.1 | 4 - 5 | 451 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SKP1| Gene Symbol: SKP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710210.3 | XP_002710256.1 | 4 - 5 | 451 | 76 | |
Set Membership: |
|