Assay Details
| Gene Symbol: MIF4GD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722950.3 | XP_002722996.2 | 207 | 95 | ||
| XM_008250964.2 | XP_008249186.1 | 408 | 95 | |||
| XM_008250965.2 | XP_008249187.1 | 267 | 95 | |||
| XM_008250967.2 | XP_008249189.1 | 201 | 95 | |||
| XM_017338929.1 | XP_017194418.1 | 1101 | 95 | |||
| XM_017338930.1 | XP_017194419.1 | 1049 | 95 | |||
| XM_017338931.1 | XP_017194420.1 | 930 | 95 | |||
| XM_017338932.1 | XP_017194421.1 | 1235 | 95 | |||
More Information
Set Membership: |
|

MIF4GD
Genomic Map