Assay Details
| Gene Symbol: IDH3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721591.3 | XP_002721637.1 | 1091 | 57 | ||
| XM_017337649.1 | XP_017193138.1 | 922 | 57 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IDH3A| Gene Symbol: IDH3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721591.3 | XP_002721637.1 | 1091 | 57 | ||
| XM_017337649.1 | XP_017193138.1 | 922 | 57 | |||
Set Membership: |
|