Assay Details
| Gene Symbol: RAB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017344012.1 | XP_017199501.1 | 5 - 6 | 598 | 84 | |
| XM_017344013.1 | XP_017199502.1 | 4 - 5 | 508 | 84 | ||
| XM_017344014.1 | XP_017199503.1 | 4 - 5 | 1112 | 84 | ||
| XM_017344015.1 | XP_017199504.1 | 5 - 6 | 1008 | 84 | ||
| XM_017344016.1 | XP_017199505.1 | 5 - 6 | 421 | 84 | ||
More Information
Set Membership: |
|

RAB12
Genomic Map