Assay Details
| Gene Symbol: TET1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008270043.2 | XP_008268265.1 | 9 - 10 | 5072 | 64 | |
| XM_017348738.1 | XP_017204227.1 | 8 - 9 | 4985 | 64 | ||
More Information
Set Membership: |
|

TET1
Genomic Map