Assay Details
| Gene Symbol: CD34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008268471.2 | XP_008266693.1 | 4 - 5 | 827 | 63 | |
| XM_008268472.2 | XP_008266694.1 | 4 - 5 | 826 | 63 | ||
More Information
Set Membership: |
|

CD34
Genomic Map