Assay Details
| Gene Symbol: CDIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722263.3 | XP_002722309.1 | 473 | 100 | ||
| XM_017338202.1 | XP_017193691.1 | 481 | 100 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDIP1| Gene Symbol: CDIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722263.3 | XP_002722309.1 | 473 | 100 | ||
| XM_017338202.1 | XP_017193691.1 | 481 | 100 | |||
Set Membership: |
|