Assay Details
| Gene Symbol: NPTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008250268.2 | XP_008248490.1 | 954 | 61 | ||
| XM_017338554.1 | XP_017194043.1 | 954 | 61 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NPTN| Gene Symbol: NPTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008250268.2 | XP_008248490.1 | 954 | 61 | ||
| XM_017338554.1 | XP_017194043.1 | 954 | 61 | |||
Set Membership: |
|