Assay Details
| Gene Symbol: ZNF232 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017349150.1 | XP_017204639.1 | 2 - 3 | 654 | 78 | |
| XM_017349151.1 | XP_017204640.1 | 2 - 3 | 722 | 78 | ||
More Information
Set Membership: |
|

ZNF232
Genomic Map