Assay Details
| Gene Symbol: TIMM10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709160.3 | XP_002709206.1 | 2 - 3 | 202 | 68 | |
| XM_008271390.2 | XP_008269612.1 | 2 - 3 | 218 | 68 | ||
| XM_008271397.2 | XP_008269619.1 | 2 - 3 | 213 | 68 | ||
| XM_008271405.2 | XP_008269627.1 | 2 - 3 | 237 | 68 | ||
More Information
Set Membership: |
|

TIMM10
Genomic Map