Assay Details
| Gene Symbol: SEPTIN11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267700.2 | XP_008265922.1 | 2 - 3 | 316 | 84 | |
| XM_008267701.2 | XP_008265923.1 | 2 - 3 | 316 | 84 | ||
| XM_008267705.2 | XP_008265927.1 | 2 - 3 | 316 | 84 | ||
| XM_008267706.2 | XP_008265928.1 | 2 - 3 | 316 | 84 | ||
| XM_008267707.2 | XP_008265929.1 | 2 - 3 | 316 | 84 | ||
| XM_008267708.2 | XP_008265930.1 | 2 - 3 | 251 | 84 | ||
| XM_017347481.1 | XP_017202970.1 | 2 - 3 | 316 | 84 | ||
| XM_017347482.1 | XP_017202971.1 | 2 - 3 | 162 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008267708.2 transcripts.
Set Membership: |
|

SEPTIN11
Genomic Map