Assay Details
| Gene Symbol: SEPTIN11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267700.2 | XP_008265922.1 | 8 - 9 | 1259 | 69 | |
| XM_008267701.2 | XP_008265923.1 | 8 - 9 | 1259 | 69 | ||
| XM_008267705.2 | XP_008265927.1 | 8 - 9 | 1259 | 69 | ||
| XM_008267706.2 | XP_008265928.1 | 8 - 9 | 1259 | 69 | ||
| XM_008267707.2 | XP_008265929.1 | 8 - 9 | 1259 | 69 | ||
| XM_008267708.2 | XP_008265930.1 | 8 - 9 | 1194 | 69 | ||
| XM_017347481.1 | XP_017202970.1 | 8 - 9 | 1259 | 69 | ||
| XM_017347482.1 | XP_017202971.1 | 8 - 9 | 1105 | 69 | ||
More Information
Set Membership: |
|

SEPTIN11
Genomic Map