Assay Details
| Gene Symbol: SEPTIN11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267706.2 | XP_008265928.1 | 9 - 10 | 1434 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SEPTIN11| Gene Symbol: SEPTIN11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267706.2 | XP_008265928.1 | 9 - 10 | 1434 | 71 | |
Set Membership: |
|