Assay Details
| Gene Symbol: LSM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716920.3 | XP_002716966.1 | 2 - 3 | 195 | 80 | |
| XM_008267381.2 | XP_008265603.1 | 5 - 6 | 582 | 80 | ||
| XM_008267382.2 | XP_008265604.1 | 5 - 6 | 539 | 80 | ||
| XM_008267383.2 | XP_008265605.1 | 4 - 5 | 379 | 80 | ||
| XM_008267384.2 | XP_008265606.1 | 3 - 4 | 296 | 80 | ||
| XM_008267385.2 | XP_008265607.1 | 3 - 4 | 277 | 80 | ||
More Information
Set Membership: |
|

LSM6
Genomic Map