Assay Details
| Gene Symbol: ANKRD46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710715.3 | XP_002710761.1 | 3 - 4 | 585 | 70 | |
| XM_008255795.2 | XP_008254017.1 | 4 - 5 | 655 | 70 | ||
More Information
Set Membership: |
|

ANKRD46
Genomic Map