Assay Details
| Gene Symbol: CTDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008252340.2 | XP_008250562.1 | 2117 | 51 | ||
| XM_017339792.1 | XP_017195281.1 | 2117 | 51 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTDP1| Gene Symbol: CTDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008252340.2 | XP_008250562.1 | 2117 | 51 | ||
| XM_017339792.1 | XP_017195281.1 | 2117 | 51 | |||
Set Membership: |
|