Assay Details
| Gene Symbol: LOC100341681 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257045.2 | XP_008255267.1 | 11 - 12 | 1372 | 98 | |
| XM_017342096.1 | XP_017197585.1 | 11 - 12 | 1384 | 98 | ||
| XM_017342098.1 | XP_017197587.1 | 10 - 11 | 1311 | 98 | ||
More Information
Set Membership: |
|

LOC100341681
Genomic Map