Assay Details
| Gene Symbol: MTSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257597.1 | XP_008255819.1 | 3 - 4 | 486 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MTSS2| Gene Symbol: MTSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257597.1 | XP_008255819.1 | 3 - 4 | 486 | 90 | |
Set Membership: |
|