Assay Details
| Gene Symbol: CNPPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712485.3 | XP_002712531.3 | 5 - 6 | 686 | 63 | |
| XM_017343138.1 | XP_017198627.1 | 6 - 7 | 624 | 63 | ||
| XM_017343139.1 | XP_017198628.1 | 5 - 6 | 632 | 63 | ||
More Information
Set Membership: |
|

CNPPD1
Genomic Map