Assay Details
| Gene Symbol: PAX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008265795.1 | XP_008264017.1 | 8 | 1891 | 65 | |
| XM_017346519.1 | XP_017202008.1 | 8 | 1888 | 65 | ||
More Information
Set Membership: |
|

PAX7
Genomic Map