Assay Details
Gene Symbol: DPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008250429.2 | XP_008248651.1 | 177 | 73 | ||
XM_008250430.2 | XP_008248652.1 | 177 | 73 | |||
XM_008250431.2 | XP_008248653.1 | 177 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
