Assay Details
Gene Symbol: ZNF146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008249563.2 | XP_008247785.1 | 271 | 118 | ||
XM_008249564.2 | XP_008247786.1 | 192 | 118 | |||
XM_017338161.1 | XP_017193650.1 | 190 | 118 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008249563.2, XM_008249564.2, XM_017338161.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
