Assay Details
| Gene Symbol: TFB1M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714950.3 | XP_002714996.1 | 2 - 3 | 379 | 57 | |
| XM_017345441.1 | XP_017200930.1 | 2 - 3 | 382 | 57 | ||
More Information
Set Membership: |
|

TFB1M
Genomic Map