Assay Details
| Gene Symbol: LOC100340267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017348347.1 | XP_017203836.1 | 9 | 1463 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100340267| Gene Symbol: LOC100340267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017348347.1 | XP_017203836.1 | 9 | 1463 | 79 | |
Set Membership: |
|