Assay Details
| Gene Symbol: DMAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249693.2 | XP_008247915.2 | 233 | 95 | ||
| XM_017338249.1 | XP_017193738.1 | 233 | 95 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DMAC2| Gene Symbol: DMAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249693.2 | XP_008247915.2 | 233 | 95 | ||
| XM_017338249.1 | XP_017193738.1 | 233 | 95 | |||
Set Membership: |
|