Assay Details
| Gene Symbol: NEDD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256929.2 | XP_008255151.1 | 2 - 3 | 218 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEDD1| Gene Symbol: NEDD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256929.2 | XP_008255151.1 | 2 - 3 | 218 | 68 | |
Set Membership: |
|