Assay Details
| Gene Symbol: LOC100346247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008251084.2 | XP_008249306.1 | 1 | 694 | 122 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100346247| Gene Symbol: LOC100346247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008251084.2 | XP_008249306.1 | 1 | 694 | 122 | |
Set Membership: |
|