Assay Details
| Gene Symbol: LOC100344248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017347354.1 | XP_017202843.1 | 4 - 5 | 1185 | 77 | |
| XM_017347355.1 | XP_017202844.1 | 4 - 5 | 1042 | 77 | ||
| XM_017347356.1 | XP_017202845.1 | 4 - 5 | 1186 | 77 | ||
More Information
Set Membership: |
|

LOC100344248
Genomic Map