Assay Details
| Gene Symbol: BOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710387.3 | XP_002710433.1 | 1 - 2 | 500 | 89 | |
| XM_017341262.1 | XP_017196751.1 | 1 - 2 | 506 | 89 | ||
More Information
Set Membership: |
|

BOD1
Genomic Map