Assay Details
| Gene Symbol: EXOSC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720605.3 | XP_002720651.1 | 988 | 69 | ||
| XM_017350168.1 | XP_017205657.1 | 940 | 69 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EXOSC8| Gene Symbol: EXOSC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720605.3 | XP_002720651.1 | 988 | 69 | ||
| XM_017350168.1 | XP_017205657.1 | 940 | 69 | |||
Set Membership: |
|