Assay Details
| Gene Symbol: CNOT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712476.3 | XP_002712522.1 | 1 - 2 | 144 | 106 | |
| XM_008259147.2 | XP_008257369.1 | 1 - 2 | 144 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002712476.3, XM_008259147.2 transcripts.
Set Membership: |
|

CNOT9
Genomic Map