Assay Details
| Gene Symbol: SIRPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256209.1 | XP_008254431.1 | 4 - 5 | 1575 | 63 | |
| XM_017341726.1 | XP_017197215.1 | 5 - 6 | 1578 | 63 | ||
| XM_017341727.1 | XP_017197216.1 | 5 - 6 | 1578 | 63 | ||
More Information
Set Membership: |
|

SIRPA
Genomic Map