Assay Details
| Gene Symbol: DCUN1D5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708545.3 | XP_002708591.1 | 8 | 2428 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DCUN1D5| Gene Symbol: DCUN1D5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708545.3 | XP_002708591.1 | 8 | 2428 | 89 | |
Set Membership: |
|