Assay Details
| Gene Symbol: KCNN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715455.3 | XP_002715501.1 | 7 - 8 | 2141 | 60 | |
| XM_008264368.2 | XP_008262590.1 | 7 - 8 | 1086 | 60 | ||
| XM_008264369.2 | XP_008262591.1 | 7 - 8 | 1116 | 60 | ||
| XM_008264370.2 | XP_008262592.1 | 7 - 8 | 1682 | 60 | ||
More Information
Set Membership: |
|

KCNN3
Genomic Map