Assay Details
| Gene Symbol: FECH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713647.3 | XP_002713693.1 | 3 - 4 | 319 | 57 | |
| XM_008261397.2 | XP_008259619.1 | 3 - 4 | 428 | 57 | ||
| XM_017344203.1 | XP_017199692.1 | 3 - 4 | 429 | 57 | ||
| XM_017344204.1 | XP_017199693.1 | 2 - 3 | 202 | 57 | ||
More Information
Set Membership: |
|

FECH
Genomic Map