Assay Details
| Gene Symbol: VIRMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710693.3 | XP_002710739.1 | 18 - 19 | 4538 | 104 | |
| XM_017341441.1 | XP_017196930.1 | 17 - 18 | 4454 | 104 | ||
More Information
Set Membership: |
|

VIRMA
Genomic Map