Assay Details
| Gene Symbol: VIRMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710693.3 | XP_002710739.1 | 2 - 3 | 332 | 107 | |
| XM_008255752.2 | XP_008253974.1 | 2 - 3 | 335 | 107 | ||
More Information
Set Membership: |
|

VIRMA
Genomic Map