Assay Details
| Gene Symbol: CNNM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710030.3 | XP_002710076.1 | 6 - 7 | 2169 | 58 | |
| XM_017339876.1 | XP_017195365.1 | 7 - 8 | 2232 | 58 | ||
More Information
Set Membership: |
|

CNNM4
Genomic Map