Assay Details
| Gene Symbol: PIR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719917.3 | XP_002719963.1 | 4 - 5 | 463 | 74 | |
| XM_017349754.1 | XP_017205243.1 | 4 - 5 | 325 | 74 | ||
More Information
Set Membership: |
|

PIR
Genomic Map