Assay Details
| Gene Symbol: METTL22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711744.3 | XP_002711790.1 | 9 - 10 | 1770 | 55 | |
| XM_008257687.2 | XP_008255909.1 | 9 - 10 | 1122 | 55 | ||
| XM_008257688.2 | XP_008255910.1 | 9 - 10 | 1119 | 55 | ||
| XM_008257690.2 | XP_008255912.1 | 8 - 9 | 993 | 55 | ||
| XM_008257691.2 | XP_008255913.1 | 7 - 8 | 936 | 55 | ||
| XM_017342440.1 | XP_017197929.1 | 10 - 11 | 1228 | 55 | ||
More Information
Set Membership: |
|

METTL22
Genomic Map